Merck
Pathogenic Streptomyces spp. Abundance Affected by Potato Cultivars.
Phytopathology (2018-04-17)
Kamrun Nahar, Claudia Goyer, Bernie J Zebarth, David L Burton, Sean Whitney
ZUSAMMENFASSUNG
Potato cultivars vary in their tolerance to common scab; however, how they affect common scab-causing Streptomyces spp. populations over time is poorly understood. This study investigated the effects of potato cultivar on pathogenic Streptomyces spp. abundance, measured using quantitative PCR, in three spatial locations in a common scab-infested field: (i) soil close to the plant (SCP); (ii) rhizosphere soil (RS); and (iii) geocaulosphere soil (GS). Two tolerant (Gold Rush, Hindenburg) and two susceptible cultivars (Green Mountain, Agria) were tested. The abundance of pathogenic Streptomyces spp. significantly increased in late August compared with other dates in RS of susceptible cultivars in both years. Abundance of pathogenic Streptomyces spp., when averaged over locations and time, was significantly greater in susceptible cultivars compared with tolerant cultivars in 2014. Principal coordinates analysis showed that SCP and RS soil properties (pH, organic carbon, and nitrogen concentrations) explained 68 and 76% of total variation in Streptomyces spp. abundance among cultivars in 2013, respectively, suggesting that cultivars influenced common scab pathogen growth conditions. The results suggested that the genetic background of potato cultivars influenced the abundance of pathogenic Streptomyces spp., with five to six times more abundant Streptomyces spp. in RS of susceptible cultivars compared with tolerant cultivars, which would result in substantially more inoculum left in the field after harvest.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
6-Carboxyfluorescein, ≥96% (HPLC)