Merck
CEP55 is a determinant of cell fate during perturbed mitosis in breast cancer.
EMBO molecular medicine (2018-08-16)
Murugan Kalimutho, Debottam Sinha, Jessie Jeffery, Katia Nones, Sriganesh Srihari, Winnie C Fernando, Pascal Hg Duijf, Claire Vennin, Prahlad Raninga, Devathri Nanayakkara, Deepak Mittal, Jodi M Saunus, Sunil R Lakhani, J Alejandro López, Kevin J Spring, Paul Timpson, Brian Gabrielli, Nicola Waddell, Kum Kum Khanna
ZUSAMMENFASSUNG
The centrosomal protein, CEP55, is a key regulator of cytokinesis, and its overexpression is linked to genomic instability, a hallmark of cancer. However, the mechanism by which it mediates genomic instability remains elusive. Here, we showed that CEP55 overexpression/knockdown impacts survival of aneuploid cells. Loss of CEP55 sensitizes breast cancer cells to anti-mitotic agents through premature CDK1/cyclin B activation and CDK1 caspase-dependent mitotic cell death. Further, we showed that CEP55 is a downstream effector of the MEK1/2-MYC axis. Blocking MEK1/2-PLK1 signaling therefore reduced outgrowth of basal-like syngeneic and human breast tumors in in vivo models. In conclusion, high CEP55 levels dictate cell fate during perturbed mitosis. Forced mitotic cell death by blocking MEK1/2-PLK1 represents a potential therapeutic strategy for MYC-CEP55-dependent basal-like, triple-negative breast cancers.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Nocodazol, ≥99% (TLC), powder
Sigma-Aldrich
RO-3306, ≥98% (HPLC)
Sigma-Aldrich
Lapatinib