Merck
  • The N-recognin UBR4 of the N-end rule pathway is targeted to and required for the biogenesis of the early endosome.
The N-recognin UBR4 of the N-end rule pathway is targeted to and required for the biogenesis of the early endosome.
Journal of cell science (2018-08-17)
Sung Tae Kim, Yoon Jee Lee, Takafumi Tasaki, Su Ran Mun, Joonsung Hwang, Min Jueng Kang, Srinivasrao Ganipisetti, Eugene C Yi, Bo Yeon Kim, Yong Tae Kwon
ZUSAMMENFASSUNG
The N-end rule pathway is a proteolytic system in which single N-terminal residues of proteins act as N-degrons. These degrons are recognized by N-recognins, facilitating substrate degradation via the ubiquitin (Ub) proteasome system (UPS) or autophagy. We have previously identified a set of N-recognins [UBR1, UBR2, UBR4 (also known as p600) and UBR5 (also known as EDD)] that bind N-degrons through their UBR boxes to promote proteolysis by the proteasome. Here, we show that the 570 kDa N-recognin UBR4 is associated with maturing endosomes through an interaction with Ca2+-bound calmodulin. The endosomal recruitment of UBR4 is essential for the biogenesis of early endosomes (EEs) and endosome-related processes, such as the trafficking of endocytosed protein cargos and degradation of extracellular cargos by endosomal hydrolases. In mouse embryos, UBR4 marks and plays a role in the endosome-lysosome pathway that mediates the heterophagic proteolysis of endocytosed maternal proteins into amino acids. By screening 9591 drugs through the DrugBank database, we identify picolinic acid as a putative ligand for UBR4 that inhibits the biogenesis of EEs. Our results suggest that UBR4 is an essential modulator in the endosome-lysosome system.This article has an associated First Person interview with the first author of the paper.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Proteasehemmer-Cocktail, for use with mammalian cell and tissue extracts, DMSO solution
Sigma-Aldrich
Poly-L-Lysin -Lösung, 0.1 % (w/v) in H2O
Sigma-Aldrich
Anti-β-Actin-Antikörper, Maus monoklonal, clone AC-15, purified from hybridoma cell culture
Sigma-Aldrich
Nocodazol, ≥99% (TLC), powder
Sigma-Aldrich
Anti-GAPDH-Antikörper, Maus monoklonal in Maus hergestellte Antikörper, clone GAPDH-71.1, purified from hybridoma cell culture
Sigma-Aldrich
Anti-LAMP1 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Latrunculin A, from sea sponge, ≥85% (HPLC), waxy solid
Sigma-Aldrich
Latrunculin A, Latrunculia magnifica, Latrunculin A, CAS 76343-93-6, is a cell-permeable marine toxin that disrupts microfilament organization in cultured cells by the formation of a 1:1 complex with monomeric G-actin (KD = 200 nM).
Sigma-Aldrich
Anti-V5 in Kaninchen hergestellte Antikörper, IgG fraction of antiserum
Sigma-Aldrich
Anti-Rab7 antibody, Mouse monoclonal, ~2 mg/mL, clone Rab7-117, purified from hybridoma cell culture
Sigma-Aldrich
W-7, Hydrochlorid – CAS 61714-27-0 – Calbiochem, A cell-permeable and reversible calmodulin antagonist that inhibits myosin light chain kinase (IC50 = 51 µM) and Ca2+-calmodulin-dependent phosphodiesterase (IC50 = 28 µM).
Sigma-Aldrich
MISSION® esiRNA, targeting human UBR4