Merck
  • Development and characterization of microencapsules containing spray dried powder obtained from Brazilian brown, green and red propolis.
Development and characterization of microencapsules containing spray dried powder obtained from Brazilian brown, green and red propolis.
Food research international (Ottawa, Ont.) (2018-05-29)
Julianna Karla Santana Andrade, Marina Denadai, George Ricardo Santana Andrade, Cristiane da Cunha Nascimento, Paula Ferreira Barbosa, Monica S Jesus, Narendra Narain
ZUSAMMENFASSUNG
The aim of this study was to obtain dried powders from the brown, green and red varieties of the Brazilian propolis by spray drying, using gum arabic and maltodextrin as encapsulating materials. Propolis microparticles were evaluated for particle size, X-Ray Diffraction (XRD), Fourier Transform Infrared Spectroscopy (FTIR), Scanning Electron Microscopy (SEM), moisture, water activity, solubility, hygroscopicity, encapsulation efficiency, total phenolic content, flavonoids and antioxidant activity (DPPH, ABTS+, FRAP, ORAC). Bioactive compounds were determined by UPHLC-QqQ-MS/MS. The results showed that the microparticles presented spherical forms, smooth surfaces, amorphous characteristics, low water activity, moisture and hygroscopicity, high solubility, good encapsulation efficiency, good antioxidant activity, although a reduction in total phenolics and flavonoids from the non-encapsulated propolis. The most important compounds in propolis powders were artepillin C, kaempferide, p-coumaric acid, luteolin, chlorogenic acid, kaempferol and caffeic acid. Spray-drying of propolis is a viable option to increase its use in food and pharmaceutical applications.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Caffeic acid phenethyl ester, ≥97% (HPLC), powder
Sigma-Aldrich
Gallussäure-ethylester, antioxidant, ≥96.0% (HPLC)
Supelco
Pinocembrin, analytical standard, ≥95.0% (HPLC), solid
Supelco
Kaempferid, analytical standard