Merck
  • Shedding Light on the Molecular Pathology of Amyloid Plaques in Transgenic Alzheimer's Disease Mice Using Multimodal MALDI Imaging Mass Spectrometry.
Shedding Light on the Molecular Pathology of Amyloid Plaques in Transgenic Alzheimer's Disease Mice Using Multimodal MALDI Imaging Mass Spectrometry.
ACS chemical neuroscience (2018-04-13)
Ibrahim Kaya, Henrik Zetterberg, Kaj Blennow, Jörg Hanrieder
ZUSAMMENFASSUNG
Senile plaques formed by aggregated amyloid β peptides are one of the major pathological hallmarks of Alzheimer's disease (AD) which have been suggested to be the primary influence triggering the AD pathogenesis and the rest of the disease process. However, neurotoxic Aβ aggregation and progression are associated with a wide range of enigmatic biochemical, biophysical and genetic processes. MALDI imaging mass spectrometry (IMS) is a label-free method to elucidate the spatial distribution patterns of intact molecules in biological tissue sections. In this communication, we utilized multimodal MALDI-IMS analysis on 18 month old transgenic AD mice (tgArcSwe) brain tissue sections to enhance molecular information correlated to individual amyloid aggregates on the very same tissue section. Dual polarity MALDI-IMS analysis of lipids on the same pixel points revealed high throughput lipid molecular information including sphingolipids, phospholipids, and lysophospholipids which can be correlated to the ion images of individual amyloid β peptide isoforms at high spatial resolutions (10 μm). Further, multivariate image analysis was applied in order to probe the multimodal MALDI-IMS data in an unbiased way which verified the correlative accumulations of lipid species with dual polarity and Aβ peptides. This was followed by the lipid fragmentation obtained directly on plaque aggregates at higher laser pulse energies which provided tandem MS information useful for structural elucidation of several lipid species. Majority of the amyloid plaque-associated alterations of lipid species are for the first time reported here. The significance of this technique is that it allows correlating the biological discussion of all detected plaque-associated molecules to the very same individual amyloid plaques which can give novel insights into the molecular pathology of even a single amyloid plaque microenvironment in a specific brain region. Therefore, this allowed us to interpret the possible roles of lipids and amyloid peptides in amyloid plaque-associated pathological events such as focal demyelination, autophagic/lysosomal dysfunction, astrogliosis, inflammation, oxidative stress, and cell death.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Yttrium-Aluminium-Oxid, nanopowder, <150 nm particle size (TEM), 99% trace metals basis