Merck
  • Injectable trace minerals (selenium, copper, zinc, and manganese) alleviate inflammation and oxidative stress during an aflatoxin challenge in lactating multiparous Holstein cows.
Injectable trace minerals (selenium, copper, zinc, and manganese) alleviate inflammation and oxidative stress during an aflatoxin challenge in lactating multiparous Holstein cows.
Journal of dairy science (2018-06-25)
R T Pate, F C Cardoso
ZUSAMMENFASSUNG
Trace minerals are vital in the antioxidant response during oxidative stress; however, limited research is available on the effects of trace mineral supplementation during an aflatoxin (AF) challenge. The objective of the study was to determine the effects of 2 subcutaneous injections of 15 mg/mL of Cu, 5 mg/mL of Se, 60 mg/mL of Zn, and 10 mg/mL of Mn (Multimin 90, Multimin North America, Fort Collins, CO) given at 1 mL/90.7 kg of average body weight in response to an AF challenge. Fifty-eight Holstein cows [body weight (mean ± SD) = 734 ± 6 0kg; days in milk = 191 ± 93] were assigned to 1 of 3 treatments in a randomized complete block design. The experimental period (63 d) was divided into an adaptation phase (d 1-56) and a measurement phase (d 57-63). From d 57 to 59, cows received an AF challenge that consisted of 100 μg of aflatoxin B1/kg of dietary dry matter intake (DMI) administered orally via balling gun. Treatments were saline injection and no AF challenge (NEG), saline injection and AF challenge (POS), and trace mineral injection and AF challenge (MM). Injections were performed subcutaneously on d 1 and 29. Milk was sampled 3 times daily from d 56 to 63, blood was sampled on d 0, 56, 60, and 63, and liver samples were taken on d 0 and 60. Two treatment orthogonal contrasts [CONT1 (NEG vs. POS) and CONT2 (POS vs. MM)] were made. Cows in NEG had lower AF excretion in milk and greater 3.5% fat-corrected milk (32.1 ± 1.37 kg/d) compared with cows in POS (28.6 ± 1.43 kg/d). Feed efficiencies (3.5% fat-corrected milk/DMI, energy-corrected milk/DMI, and milk/DMI) were greater for cows in NEG (1.42 ± 0.07, 1.46 ± 0.07, and 1.45 ± 0.07, respectively) than cows in POS (1.16 ± 0.08, 1.18 ± 0.08, and 1.22 ± 0.07, respectively). Cows in POS had greater milk urea nitrogen and blood urea nitrogen than cows in MM. Liver concentrations of Se and Fe were greater for cows in MM compared with cows in POS. Cows in MM tended to have greater plasma glutathione peroxidase activity compared with cows in POS. An upregulation of liver GPX1 was observed for cows in POS compared with cows in MM. In conclusion, subcutaneous injection of trace minerals maintained an adequate antioxidant response when an AF challenge was present.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
S-Nitroso-N-acetyl-DL-penicillamin, ≥97%, powder