Merck
  • Effects of a high-sodium/low-potassium diet on renal calcium, magnesium, and phosphate handling.
Effects of a high-sodium/low-potassium diet on renal calcium, magnesium, and phosphate handling.
American journal of physiology. Renal physiology (2018-01-24)
Jenny van der Wijst, Omar A Z Tutakhel, Caro Bos, Alexander H J Danser, Ewout J Hoorn, Joost G J Hoenderop, René J M Bindels
ZUSAMMENFASSUNG
The distal convoluted tubule (DCT) of the kidney plays an important role in blood pressure regulation by modulating Na+ reabsorption via the Na+-Cl- cotransporter (NCC). A diet containing high salt (NaCl) and low K+ activates NCC, thereby causing Na+ retention and a rise in blood pressure. Since high blood pressure, hypertension, is associated with changes in serum calcium (Ca2+) and magnesium (Mg2+) levels, we hypothesized that dietary Na+ and K+ intake affects Ca2+ and Mg2+ transport in the DCT. Therefore, the present study aimed to investigate the effect of a high-Na+/low-K+ diet on renal Ca2+ and Mg2+ handling. Mice were divided in four groups and fed a normal-Na+/normal-K+, normal-Na+/low-K+, high-Na+/normal-K+, or high-Na+/low-K+ diet for 4 days. Serum and urine were collected for electrolyte and hormone analysis. Gene and protein expression of electrolyte transporters were assessed in kidney and intestine by qPCR and immunoblotting. Whereas Mg2+ homeostasis was not affected, the mice had elevated urinary Ca2+ and phosphate (Pi) excretion upon high Na+ intake, as well as significantly lower serum Ca2+ levels in the high-Na+/low-K+ group. Alterations in the gene and protein expression of players involved in Ca2+ and Pi transport indicate that reabsorption in the proximal tubular and TAL is affected, while inducing a compensatory response in the DCT. These effects may contribute to the negative health impact of a high-salt diet, including kidney stone formation, chronic kidney disease, and loss of bone mineral density.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Monoklonales Anti-β-Aktin in Maus hergestellte Antikörper, clone AC-15, ascites fluid
Sigma-Aldrich
Monoklonaler Anti-Calbindin-D-28K-Antikörper in Maus hergestellte Antikörper, clone CB-955, ascites fluid
Sigma-Aldrich
DL-Glyceraldehyd-3-phosphat -Lösung, 45-55 mg/mL in H2O
Sigma-Aldrich
Anti-Kaninchen-IgG-(Gesamtmolekül)-Peroxidase in Ziege hergestellte Antikörper, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Anti-Thiazid-sensitiver NaCl-Cotransporter-Antikörper, Chemicon®, from rabbit