Merck
Novel Fubinaca/Rimonabant hybrids as endocannabinoid system modulators.
Amino acids (2018-08-27)
Azzurra Stefanucci, Giorgia Macedonio, Szabolcs Dvorácskó, Csaba Tömböly, Adriano Mollica
ZUSAMMENFASSUNG
The discovery of novel modulators of the cannabinoid system is a current topic in medicinal chemistry. In this paper, we report nine novel carboxamides designed as hybrids of Fubinaca family compounds and Rimonabant. These hybrids were obtained by linking the 1-benzyl-2,5-dichloroindazole-3-carboxylic acid to different amino acids bearing a hydrophobic side chain and three different C-terminus. The new chemical entities were tested in vitro to evaluate their bioactivity by means of receptor binding assays and [35S]GTPγS stimulation assays to reveal their affinity and potency. We found that all compounds were able to bind to the cannabinoid receptors in the low nanomolar range with a marked selectivity towards the CB1 cannabinoid receptor. Some of them are full agonists, whereas the others act as partial agonists. These molecules could be potentially used as anti-obesity agents, antiemetic and analgesics.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Lonidamine, mitochondrial hexokinase inhibitor