Merck
The MALDI-TOF E2/E3 Ligase Assay as Universal Tool for Drug Discovery in the Ubiquitin Pathway.
Cell chemical biology (2018-07-19)
Virginia De Cesare, Clare Johnson, Victoria Barlow, James Hastie, Axel Knebel, Matthias Trost
ZUSAMMENFASSUNG
Due to their role in many diseases, enzymes of the ubiquitin system have recently become interesting drug targets. Despite efforts, primary screenings of compound libraries targeting E2 enzymes and E3 ligases have been strongly limited by the lack of robust and fast high-throughput assays. Here we report a label-free high-throughput screening assay for ubiquitin E2 conjugating enzymes and E3 ligases based on MALDI-TOF mass spectrometry. The MALDI-TOF E2/E3 assay allows testing E2 enzymes and E3 ligases for their ubiquitin transfer activity, identifying E2/E3 active pairs, inhibitor potency and specificity and screening compound libraries in vitro without chemical or fluorescent probes. We demonstrate that the MALDI-TOF E2/E3 assay is a universal tool for drug discovery screening in the ubiquitin pathway as it is suitable for working with all E3 ligase families and requires a reduced amount of reagents, compared with standard biochemical assays.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Dihydroxyacetonphosphat Dilithiumsalz, ≥93% dry basis (enzymatic)
Sigma-Aldrich
Bay 11-7082, ≥98% (HPLC), powder
Sigma-Aldrich
Gliotoxin aus Gliocladium fimbriatum
Sigma-Aldrich
DUB-Inhibitor V, PR-619, Calbiochem, The DUB Inhibitor V, PR-619 controls the biological activity of DUB. This small molecule/inhibitor is primarily used for Protease Inhibitors applications.
Sigma-Aldrich
Bendamustine hydrochloride hydrate, ≥98% (HPLC)
Sigma-Aldrich
PYR-41, ≥98% (HPLC), powder