Merck
  • Prenatal influenza vaccination rescues impairments of social behavior and lamination in a mouse model of autism.
Prenatal influenza vaccination rescues impairments of social behavior and lamination in a mouse model of autism.
Journal of neuroinflammation (2018-08-15)
Yingying Wu, Fangfang Qi, Dan Song, Zitian He, Zejie Zuo, Yunjie Yang, Qiongliang Liu, Saisai Hu, Xiao Wang, Xiaona Zheng, Junhua Yang, Qunfang Yuan, Juntao Zou, Kaihua Guo, Zhibin Yao
ZUSAMMENFASSUNG
Prenatal infection is a substantial risk factor for neurodevelopmental disorders such as autism in offspring. We have previously reported that influenza vaccination (VAC) during early pregnancy contributes to neurogenesis and behavioral function in offspring. Here, we probe the efficacy of VAC pretreatment on autism-like behaviors in a lipopolysaccharide (LPS)-induced maternal immune activation (MIA) mouse model. We show that VAC improves abnormal fetal brain cytoarchitecture and lamination, an effect associated with promotion of intermediate progenitor cell differentiation in MIA fetal brain. These beneficial effects are sufficient to prevent social deficits in adult MIA offspring. Furthermore, whole-genome analysis suggests a strong interaction between Ikzf1 (IKAROS family zinc-finger 1) and neuronal differentiation. Intriguingly, VAC rescues excessive microglial Ikzf1 expression and attenuates microglial inflammatory responses in the MIA fetal brain. Our study implies that a preprocessed influenza vaccination prevents maternal bacterial infection from causing neocortical lamination impairments and autism-related behaviors in offspring.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Lipopolysaccharide aus Escherichia coli O111:B4, purified by phenol extraction
Sigma-Aldrich
Triton X-100, laboratory grade
Sigma-Aldrich
Anti-NeuN-Antikörper, Klon A60, clone A60, Chemicon®, from mouse