Merck
  • The Endosomal Protein CEMIP Links WNT Signaling to MEK1-ERK1/2 Activation in Selumetinib-Resistant Intestinal Organoids.
The Endosomal Protein CEMIP Links WNT Signaling to MEK1-ERK1/2 Activation in Selumetinib-Resistant Intestinal Organoids.
Cancer research (2018-06-20)
Hong Quan Duong, Ivan Nemazanyy, Florian Rambow, Seng Chuan Tang, Sylvain Delaunay, Lars Tharun, Alexandra Florin, Reinhard Büttner, Daniel Vandaele, Pierre Close, Jean-Christophe Marine, Kateryna Shostak, Alain Chariot
ZUSAMMENFASSUNG
MAPK signaling pathways are constitutively active in colon cancer and also promote acquired resistance to MEK1 inhibition. Here, we demonstrate that BRAFV600E -mutated colorectal cancers acquire resistance to MEK1 inhibition by inducing expression of the scaffold protein CEMIP through a β-catenin- and FRA-1-dependent pathway. CEMIP was found in endosomes and bound MEK1 to sustain ERK1/2 activation in MEK1 inhibitor-resistant BRAFV600E-mutated colorectal cancers. The CEMIP-dependent pathway maintained c-Myc protein levels through ERK1/2 and provided metabolic advantage in resistant cells, potentially by sustaining amino acids synthesis. CEMIP silencing circumvented resistance to MEK1 inhibition, partly, through a decrease of both ERK1/2 signaling and c-Myc. Together, our data identify a cross-talk between Wnt and MAPK signaling cascades, which involves CEMIP. Activation of this pathway promotes survival by potentially regulating levels of specific amino acids via a Myc-associated cascade. Targeting this node may provide a promising avenue for treatment of colon cancers that have acquired resistance to targeted therapies.Significance: MEK1 inhibitor-resistant colorectal cancer relies on the scaffold and endosomal protein CEMIP to maintain ERK1/2 signaling and Myc-driven transcription. Cancer Res; 78(16); 4533-48. ©2018 AACR.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
L-(−)-Dithiothreitol, ≥95% (titration)
Sigma-Aldrich
PNU-74654, ≥98% (HPLC), solid