Merck
A systems-level framework for drug discovery identifies Csf1R as an anti-epileptic drug target.
Nature communications (2018-09-05)
Prashant K Srivastava, Jonathan van Eyll, Patrice Godard, Manuela Mazzuferi, Andree Delahaye-Duriez, Juliette Van Steenwinckel, Pierre Gressens, Benedicte Danis, Catherine Vandenplas, Patrik Foerch, Karine Leclercq, Georges Mairet-Coello, Alvaro Cardenas, Frederic Vanclef, Liisi Laaniste, Isabelle Niespodziany, James Keaney, Julien Gasser, Gaelle Gillet, Kirill Shkura, Seon-Ah Chong, Jacques Behmoaras, Irena Kadiu, Enrico Petretto, Rafal M Kaminski, Michael R Johnson
ZUSAMMENFASSUNG
The identification of drug targets is highly challenging, particularly for diseases of the brain. To address this problem, we developed and experimentally validated a general computational framework for drug target discovery that combines gene regulatory information with causal reasoning ("Causal Reasoning Analytical Framework for Target discovery"-CRAFT). Using a systems genetics approach and starting from gene expression data from the target tissue, CRAFT provides a predictive framework for identifying cell membrane receptors with a direction-specified influence over disease-related gene expression profiles. As proof of concept, we applied CRAFT to epilepsy and predicted the tyrosine kinase receptor Csf1R as a potential therapeutic target. The predicted effect of Csf1R blockade in attenuating epilepsy seizures was validated in three pre-clinical models of epilepsy. These results highlight CRAFT as a systems-level framework for target discovery and suggest Csf1R blockade as a novel therapeutic strategy in epilepsy. CRAFT is applicable to disease settings other than epilepsy.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Kynurensäure, ≥98%