Merck
  • MALDI-TOF for the rapid detection of carbapenemase-producing Enterobacteriaceae: comparison of the commercialized MBT STAR®-Carba IVD Kit with two in-house MALDI-TOF techniques and the RAPIDEC® CARBA NP.
MALDI-TOF for the rapid detection of carbapenemase-producing Enterobacteriaceae: comparison of the commercialized MBT STAR®-Carba IVD Kit with two in-house MALDI-TOF techniques and the RAPIDEC® CARBA NP.
The Journal of antimicrobial chemotherapy (2018-06-14)
Laurent Dortet, Didier Tandé, Dominique de Briel, Sandrine Bernabeu, Camille Lasserre, Guillaume Gregorowicz, Agnès B Jousset, Thierry Naas
ZUSAMMENFASSUNG
There is an urgent need for accurate and fast diagnostic tests to identify carbapenemase-producing bacteria. Here, we have evaluated three MALDI-TOF-based techniques to detect carbapenemase-producing Enterobacteriaceae (CPE) from cultured colonies. The performance of three MALDI-TOF-based techniques, including the commercialized MBT STAR®-Carba IVD Kit (Bruker Daltonics) and two in-house protocols performed on the Microflex LT Biotyper (Bruker Daltonics) and the VITEK® MS Plus (bioMérieux), were compared with those of the RAPIDEC® CARBA NP (bioMérieux). A collection of 175 isolates including 120 carbapenemase producers and 55 non-carbapenemase producers was tested. Samples were tested blind in the three participating centres. The repeatability of the MBT STAR®-Carba IVD Kit was also evaluated. The three MALDI-TOF techniques possess sensitivities ranging from 95% to 100% and specificities from 98.2% to 100% compared with 99.2% and 100%, respectively, for the RAPIDEC® CARBA NP. The MBT STAR®-Carba IVD Kit gave highly reproducible results and is the only technique able to provide a concomitant identification of the bacterial isolate. The three MALDI-TOF techniques possess a fast turnaround time (less than 1.5 h). Overall, MALDI-TOF is a reliable technique for the rapid detection of CPE from cultured colonies. MBT STAR®-Carba IVD Kit, the only commercially available assay, could easily be implemented in a clinical microbiology laboratory if it is already equipped with a Microflex LT Biotyper mass spectrometer.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Supelco
Imipenem, Pharmaceutical Secondary Standard; Certified Reference Material