Merck
  • Antibacterial, anti-inflammatory, and bone-regenerative dual-drug-loaded calcium phosphate nanocarriers-in vitro and in vivo studies.
Antibacterial, anti-inflammatory, and bone-regenerative dual-drug-loaded calcium phosphate nanocarriers-in vitro and in vivo studies.
Drug delivery and translational research (2018-05-03)
K Madhumathi, Y Rubaiya, Mukesh Doble, R Venkateswari, T S Sampath Kumar
ZUSAMMENFASSUNG
A dual local drug delivery system (DDS) composed of calcium phosphate bioceramic nanocarriers aimed at treating the antibacterial, anti-inflammatory, and bone-regenerative aspects of periodontitis has been developed. Calcium-deficient hydroxyapatite (CDHA, Ca/P = 1.61) and tricalcium phosphate (β-TCP) were prepared by microwave-accelerated wet chemical synthesis method. The phase purity of the nanocarriers was confirmed by x-ray diffraction (XRD) and Fourier transform infrared spectroscopy (FT-IR), while the transmission electron microscopy (TEM) confirmed their nanosized morphology. CDHA was selected as carrier for the antibiotic (tetracycline) while TCP was chosen as the anti-inflammatory drug (ibuprofen) carrier. Combined drug release profile was studied in vitro from CDHA/TCP (CTP) system and compared with a HA/TCP (BCP) biphasic system. The tetracycline and ibuprofen release rate was 71 and 23% from CTP system as compared to 63 and 20% from BCP system. CTP system also showed a more controlled drug release profile compared to BCP system. Modeling of drug release kinetics from CTP system indicated that the release follows Higuchi model with a non-typical Fickian diffusion profile. In vitro biological studies showed the CTP system to be biocompatible with significant antibacterial and anti-inflammatory activity. In vivo implantation studies on rat cranial defects showed greater bone healing and new bone formation in the drug-loaded CTP system compared to control (no carrier) at the end of 12 weeks. The in vitro and in vivo results suggest that the combined drug delivery platform can provide a comprehensive management for all bone infections requiring multi-drug therapy.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Supelco
Calcium Standard für AAS, analytical standard, 1.000 g/L Ca2+ in hydrochloric acid, traceable to BAM