Merck
  • Pt(II)-activated coupling of aminoethylferrocene with benzonitrile. A facile access route to a new redox-active bis(ferrocenyl-amidine) anion sensor.
Pt(II)-activated coupling of aminoethylferrocene with benzonitrile. A facile access route to a new redox-active bis(ferrocenyl-amidine) anion sensor.
Chemical communications (Cambridge, England) (2011-08-17)
Daniel Nieto, Ana Ma González-Vadillo, Sonia Bruña, César J Pastor, Angel E Kaifer, Isabel Cuadrado
ZUSAMMENFASSUNG
A new heterometallic electroactive compound having the ferrocenyl moiety attached to the platinum atom through an amidine functionality has been prepared via Pt(II)-mediated nucleophilic addition of β-aminoethylferrocene to a benzonitrile ligand. This neutral receptor shows remarkable electrochemical anion-sensing redox response to H(2)PO(4)(-).
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Benzonitril, ReagentPlus®, 99%