- Synthesis of omega-hydroxy carboxylic acids and alpha,omega-dimethyl ketones using alpha,omega-diols as alkylating agents.
Synthesis of omega-hydroxy carboxylic acids and alpha,omega-dimethyl ketones using alpha,omega-diols as alkylating agents.
The Journal of organic chemistry (2010-02-04)
Yosuke Iuchi, Megumi Hyotanishi, Brittany E Miller, Kensaku Maeda, Yasushi Obora, Yasutaka Ishii
PMID20121069
ZUSAMMENFASSUNG
Synthesis of omega-hydroxy carboxylic acids and alpha,omega-dimethyl diketones was successfully achieved by using alpha,omega-diols as alkylating agents under the influence of an iridium catalyst. For example, the alkylation of butyl cyanoacetate with 1,13-tridecanediol in the presence of [IrCl(cod)](2) or [IrCl(coe)(2)](2) gave rise to butyl 2-cyano-15-hydroxypentadecanoate in good yield which is easily converted to cyclopentadecanolide (CPDL). In addition, the alkylation of acetone with 1,10-decanediol in the presence of [IrCl(cod)](2) and KOH resulted in an important muscone precursor, 2,15-hexadecanedione (HDDO), in good yield.