Merck
  • Scinderin-knockdown inhibits proliferation and promotes apoptosis in human breast carcinoma cells.
Scinderin-knockdown inhibits proliferation and promotes apoptosis in human breast carcinoma cells.
Oncology letters (2018-08-22)
Wenjing Jian, Xiaoli Zhang, Jiguo Wang, Yunlong Liu, Chuting Hu, Xianming Wang, Renbin Liu
ZUSAMMENFASSUNG
Previous studies have reported that scinderin (SCIN) affects multiple cellular processes, including proliferation, migration and differentiation in cancer. However, the specific role of SCIN in breast cancer (BC) cells is unknown. Immunohistochemistry was used to investigate SCIN expression in 46 BC and 21 mammary fibroadenoma or fibroadenomatoid hyperplasia tissue samples. SCIN expression was ablated in MDA-MB-231 and T-47D cells using lentivirus-mediated small interfering RNA technology. Cell proliferation was tested using Celigo and 3-(4,5-dimethylthiazol-2-yl)-2,5-diphenyltetrazolium bromide assays. Cell apoptosis was analyzed by measuring Caspase 3/7 activity and annexin-V staining. The results of the present study demonstrated that SCIN expression was elevated in BC tissues compared with mammary fibroadenoma or fibroadenomatoid hyperplasia tissues. Specifically, higher SCIN expression was observed in Ki-67-positive BC tissues (78.6%) compared with Ki-67-negative BC tissues. Furthermore, knockdown of SCIN expression in the BC cell lines significantly suppressed cell proliferation and induced apoptosis. The data presented in the present study indicate that SCIN serves an important role in the development of breast cancer.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Anti-SCIN antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, Ab1