Merck
Deficiency of PRKD2 triggers hyperinsulinemia and metabolic disorders.
Nature communications (2018-05-24)
Yao Xiao, Can Wang, Jia-Yu Chen, Fujian Lu, Jue Wang, Ning Hou, Xiaomin Hu, Fanxin Zeng, Dongwei Ma, Xueting Sun, Yi Ding, Yan Zhang, Wen Zheng, Yuli Liu, Haibao Shang, Wenzhen Zhu, Chensheng Han, Yulin Zhang, Kunfu Ouyang, Liangyi Chen, Ju Chen, Rui-Ping Xiao, Chuan-Yun Li, Xiuqin Zhang
ZUSAMMENFASSUNG
Hyperinsulinemia is the earliest symptom of insulin resistance (IR), but a causal relationship between the two remains to be established. Here we show that a protein kinase D2 (PRKD2) nonsense mutation (K410X) in two rhesus monkeys with extreme hyperinsulinemia along with IR and metabolic defects by using extreme phenotype sampling and deep sequencing analyses. This mutation reduces PRKD2 at both the mRNA and the protein levels. Taking advantage of a PRKD2-KO mouse model, we demonstrate that PRKD2 deletion triggers hyperinsulinemia which precedes to IR and metabolic disorders in the PRKD2 ablation mice. PRKD2 deficiency promotes β-cell insulin secretion by increasing the expression and activity of L-type Ca2+ channels and subsequently augmenting high glucose- and membrane depolarization-induced Ca2+ influx. Altogether, these results indicate that down-regulation of PRKD2 is involved in the pathogenesis of hyperinsulinemia which, in turn, results in IR and metabolic disorders.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Monoklonaler Anti-α-Tubulin-Antikörper in Maus hergestellte Antikörper, ascites fluid, clone B-5-1-2