Merck
Molecular basis for disassembly of an importin:ribosomal protein complex by the escortin Tsr2.
Nature communications (2018-09-12)
Sabina Schütz, Erich Michel, Fred F Damberger, Michaela Oplová, Cohue Peña, Alexander Leitner, Ruedi Aebersold, Frederic H-T Allain, Vikram Govind Panse
ZUSAMMENFASSUNG
Disordered extensions at the termini and short internal insertions distinguish eukaryotic ribosomal proteins (r-proteins) from their anucleated archaeal counterparts. Here, we report an NMR structure of such a eukaryotic-specific segment (ESS) in the r-protein eS26 in complex with the escortin Tsr2. The structure reveals how ESS attracts Tsr2 specifically to importin:eS26 complexes entering the nucleus in order to trigger non-canonical RanGTP-independent disassembly. Tsr2 then sequesters the released eS26 and prevents rebinding to the importin, providing an alternative allosteric mechanism to terminate the process of nuclear import. Notably, a Diamond-Blackfan anemia-associated Tsr2 mutant protein is impaired in binding to ESS, unveiling a critical role for this interaction in human hematopoiesis. We propose that eS26-ESS and Tsr2 are components of a nuclear sorting system that co-evolved with the emergence of the nucleocytoplasmic barrier and transport carriers.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Monoklonaler ANTI-FLAG® M2-Antikörper in Maus hergestellte Antikörper, clone M2, purified immunoglobulin (Purified IgG1 subclass), buffered aqueous solution (10 mM sodium phosphate, 150 mM NaCl, pH 7.4, containing 0.02% sodium azide)
Sigma-Aldrich
Anti-Kaninchen-IgG-(Gesamtmolekül)-Peroxidase in Ziege hergestellte Antikörper, affinity isolated antibody
Sigma-Aldrich
Anti-Maus-IgG (Gesamtmolekül)-Peroxidase in Ziege hergestellte Antikörper, affinity isolated antibody, buffered aqueous solution