Merck
The FAK inhibitor BI 853520 exerts anti-tumor effects in breast cancer.
Oncogenesis (2018-09-22)
Stefanie Tiede, Nathalie Meyer-Schaller, Ravi Kiran Reddy Kalathur, Robert Ivanek, Ernesta Fagiani, Philip Schmassmann, Patrick Stillhard, Simon Häfliger, Norbert Kraut, Norbert Schweifer, Irene C Waizenegger, Ruben Bill, Gerhard Christofori
ZUSAMMENFASSUNG
Focal adhesion kinase (FAK) is a cytoplasmic tyrosine kinase that regulates a plethora of downstream signaling pathways essential for cell migration, proliferation and death, processes that are exploited by cancer cells during malignant progression. These well-established tumorigenic activities, together with its high expression and activity in different cancer types, highlight FAK as an attractive target for cancer therapy. We have assessed and characterized the therapeutic potential and the biological effects of BI 853520, a novel small chemical inhibitor of FAK, in several preclinical mouse models of breast cancer. Treatment with BI 853520 elicits a significant reduction in primary tumor growth caused by an anti-proliferative activity by BI 853520. In contrast, BI 853520 exerts effects with varying degrees of robustness on the different stages of the metastatic cascade. Together, the data demonstrate that the repression of FAK activity by the specific FAK inhibitor BI 853520 offers a promising anti-proliferative approach for cancer therapy.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Anti-Phospho-Histon H3 (Ser10)-Antikörper, Mitosemarker, Upstate®, from rabbit
Sigma-Aldrich
Anti-GAPDH-Antikörper, Maus monoklonal in Maus hergestellte Antikörper, clone GAPDH-71.1, purified from hybridoma cell culture
Sigma-Aldrich
Anti-FAK-Antikörper, Klon 4.47, clone 4.47, Upstate®, from mouse
Sigma-Aldrich
Monoclonal Anti-Vimentin antibody produced in mouse, clone LN-6, ascites fluid