Merck
  • Suboptimal Level of Bone-Forming Cells in Advanced Cirrhosis are Associated with Hepatic Osteodystrophy.
Suboptimal Level of Bone-Forming Cells in Advanced Cirrhosis are Associated with Hepatic Osteodystrophy.
Hepatology communications (2018-09-12)
Chhagan Bihari, Deepika Lal, Monika Thakur, Sukriti Sukriti, Dhananjay Mathur, Anupama G Patil, Lovkesh Anand, Guresh Kumar, Shvetank Sharma, Shalini Thapar, Apurba Rajbongshi, Archana Rastogi, Anupam Kumar, Shiv K Sarin
ZUSAMMENFASSUNG
Bone loss is common in advanced cirrhosis, although the precise mechanisms underlying bone loss in cirrhosis are unknown. We studied the profile and functionality of bone-forming cells and bone-building proteins in bone marrow (BM) of individuals with cirrhosis (n = 61) and individuals without cirrhosis as normal controls (n = 50). We also performed dual energy X-ray absorptiometry for clinical correlation. BM mesenchymal cells (MSCs) were analyzed for colony-forming units-fibroblasts and their osteogenic (fibronectin-1 [FN1], insulin-like growth factor binding protein 3 [IGFBP3], collagen type 1 alpha 1 chain [COL1A1], runt-related transcription factor 2 [RUNX2], and alkaline phosphatase, liver [ALPL]) and adipogenic ( adiponectin, C1Q, and collagen domain containing [ADIPOQ], peroxisome proliferator-activated receptor gamma [PPARγ], and fatty acid binding protein 4 [FABP4]) potentials. Colony-forming units-fibroblasts were lower in patients with cirrhosis (P = 0.002) than in controls. Cirrhotic BM-MSCs showed >2-fold decrease in osteogenic markers. Compared to controls, patients with cirrhosis showed fewer osteocytes (P = 0.05), osteoblasts, chondroblasts, osteocalcin-positive (osteocalcin+) area, clusters of differentiation (CD)169+ macrophages (P < 0.001, each), and nestin+ MSCs (P = 0.001); this was more apparent in Child-Turcotte-Pugh (CTP) class C than A (P < 0.001). Multivariate logistic regression showed low nestin+ MSCs (P = 0.004) as a predictor of bone loss. Bone-resolving osteoclasts were comparable among CTP groups, but >2-fold decreased anti-osteoclastic and increased pro-osteoclastic factors were noted in patients with CTP C compared to CTP A. Bone-building proteins (osteocalcin [P = 0.008], osteonectin [P < 0.001], and bone morphogenic protein 2 [P = 0.001]) were decreased while anti-bone repair factors (fibroblast growth factor 23 [P = 0.015] and dipeptidyl peptidase 4 [P < 0.001]) were increased in BM and peripheral blood; this was more apparent in advanced cirrhosis. The dual energy X-ray absorptiometry scan T score significantly correlated with the population of osteoblasts, osteocytes, MSCs, and CD169+ macrophages. Conclusion: Osteoprogenitor cells are substantially reduced in patients with cirrhosis and more so in advanced disease. Additionally, increased anti-bone repair proteins enhance the ineffective bone repair and development of osteoporosis in cirrhosis. Hepatology Communications 2018;0:0-0).
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Anti-Nestin antibody produced in rabbit, ~1 mg/mL, affinity isolated antibody, buffered aqueous solution