Merck
  • Immunoprecipitation and mass spectrometry defines an extensive RBM45 protein-protein interaction network.
Immunoprecipitation and mass spectrometry defines an extensive RBM45 protein-protein interaction network.
Brain research (2016-03-17)
Yang Li, Mahlon Collins, Jiyan An, Rachel Geiser, Tony Tegeler, Kristine Tsantilas, Krystine Garcia, Patrick Pirrotte, Robert Bowser
ZUSAMMENFASSUNG
The pathological accumulation of RNA-binding proteins (RBPs) within inclusion bodies is a hallmark of amyotrophic lateral sclerosis (ALS) and frontotemporal lobar degeneration (FTLD). RBP aggregation results in both toxic gain and loss of normal function. Determining the protein binding partners and normal functions of disease-associated RBPs is necessary to fully understand molecular mechanisms of RBPs in disease. Herein, we characterized the protein-protein interactions (PPIs) of RBM45, a RBP that localizes to inclusions in ALS/FTLD. Using immunoprecipitation coupled to mass spectrometry (IP-MS), we identified 132 proteins that specifically interact with RBM45 within HEK293 cells. Select PPIs were validated by immunoblot and immunocytochemistry, demonstrating that RBM45 associates with a number of other RBPs primarily via RNA-dependent interactions in the nucleus. Analysis of the biological processes and pathways associated with RBM45-interacting proteins indicates enrichment for nuclear RNA processing/splicing via association with hnRNP proteins and cytoplasmic RNA translation via eiF2 and eiF4 pathways. Moreover, several other ALS-linked RBPs, including TDP-43, FUS, Matrin-3, and hnRNP-A1, interact with RBM45, consistent with prior observations of these proteins within intracellular inclusions in ALS/FTLD. Taken together, our results define a PPI network for RBM45, suggest novel functions for this protein, and provide new insights into the contributions of RBM45 to neurodegeneration in ALS/FTLD. This article is part of a Special Issue entitled SI:RNA Metabolism in Disease.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Proteasehemmer-Cocktail, for use with mammalian cell and tissue extracts, DMSO solution
Millipore
ANTI-FLAG® M2-Affinitätsgel, purified immunoglobulin, buffered aqueous glycerol solution
Sigma-Aldrich
Monoklonaler ANTI-FLAG® M2-Antikörper in Maus hergestellte Antikörper, clone M2, purified immunoglobulin (Purified IgG1 subclass), buffered aqueous solution (10 mM sodium phosphate, 150 mM NaCl, pH 7.4, containing 0.02% sodium azide)
Millipore
Monoklonale Anti-HA−Agarose in Maus hergestellte Antikörper, clone HA-7, purified immunoglobulin, PBS suspension
Sigma-Aldrich
IgG aus Mausserum, reagent grade, ≥95% (SDS-PAGE), lyophilized powder
Sigma-Aldrich
IgG aus Kaninchenserum, reagent grade, ≥95% (SDS-PAGE), essentially salt-free, lyophilized powder
Sigma-Aldrich
Maus-IgG−Agarose, (Suspension in 0.5 M NaCl containing preservative.)
Millipore
Phosphatase-Inhibitor-Cocktail-Set V, 50X, Phosphatase Inhibitor Cocktail Set V is a 50X cocktail of four serine/threonine and tyrosine phosphatase inhibitors.
Sigma-Aldrich
Anti-SMN antibody, Mouse monoclonal, clone 2B1, purified from hybridoma cell culture