Merck
APOL1 risk allele RNA contributes to renal toxicity by activating protein kinase R.
Communications biology (2018-11-13)
Koji Okamoto, Jason W Rausch, Hidefumi Wakashin, Yulong Fu, Joon-Yong Chung, Patrick D Dummer, Myung K Shin, Preeti Chandra, Kosuke Suzuki, Shashi Shrivastav, Avi Z Rosenberg, Stephen M Hewitt, Patricio E Ray, Eisei Noiri, Stuart F J Le Grice, Maarten Hoek, Zhe Han, Cheryl A Winkler, Jeffrey B Kopp
ZUSAMMENFASSUNG
APOL1 risk alleles associate with chronic kidney disease in African Americans, but the mechanisms remain to be fully understood. We show that APOL1 risk alleles activate protein kinase R (PKR) in cultured cells and transgenic mice. This effect is preserved when a premature stop codon is introduced to APOL1 risk alleles, suggesting that APOL1 RNA but not protein is required for the effect. Podocyte expression of APOL1 risk allele RNA, but not protein, in transgenic mice induces glomerular injury and proteinuria. Structural analysis of the APOL1 RNA shows that the risk variants possess secondary structure serving as a scaffold for tandem PKR binding and activation. These findings provide a mechanism by which APOL1 variants damage podocytes and suggest novel therapeutic strategies.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Anti-APOL1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-HRI-Antikörper, serum, Upstate®