Merck
PKCε as a novel promoter of skeletal muscle differentiation and regeneration.
Experimental cell research (2015-10-04)
D Di Marcantonio, D Galli, C Carubbi, G Gobbi, V Queirolo, S Martini, S Merighi, M Vaccarezza, N Maffulli, S M Sykes, M Vitale, P Mirandola
ZUSAMMENFASSUNG
Satellite cells are muscle resident stem cells and are responsible for muscle regeneration. In this study we investigate the involvement of PKCε during muscle stem cell differentiation in vitro and in vivo. Here, we describe the identification of a previously unrecognized role for the PKCε-HMGA1 signaling axis in myoblast differentiation and regeneration processes. PKCε expression was modulated in the C2C12 cell line and primary murine satellite cells in vitro, as well as in an in vivo model of muscle regeneration. Immunohistochemistry and immunofluorescence, RT-PCR and shRNA silencing techniques were used to determine the role of PKCε and HMGA1 in myogenic differentiation. PKCε expression increases and subsequently re-localizes to the nucleus during skeletal muscle cell differentiation. In the nucleus, PKCε blocks Hmga1 expression to promote Myogenin and Mrf4 accumulation and myoblast formation. Following in vivo muscle injury, PKCε accumulates in regenerating, centrally-nucleated myofibers. Pharmacological inhibition of PKCε impairs the expression of two crucial markers of muscle differentiation, namely MyoD and Myogenin, during injury induced muscle regeneration. This work identifies the PKCε-HMGA1 signaling axis as a positive regulator of skeletal muscle differentiation.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Anti-Glyceraldehyd-3-phosphat-dehydrogenase-Antikörper, Klon 6C5, clone 6C5, Chemicon®, from mouse
Sigma-Aldrich
Monoclonal Anti-Heat Shock Protein 70 antibody produced in mouse, clone BRM-22, ascites fluid
Sigma-Aldrich
Anti-PKCε-Antikörper, Upstate®, from rabbit