Merck
  • Dihomo-γ-linolenic acid inhibits growth of xenograft tumors in mice bearing human pancreatic cancer cells (BxPC-3) transfected with delta-5-desaturase shRNA.
Dihomo-γ-linolenic acid inhibits growth of xenograft tumors in mice bearing human pancreatic cancer cells (BxPC-3) transfected with delta-5-desaturase shRNA.
Redox biology (2018-11-02)
Xiaoyu Yang, Yi Xu, Di Gao, Liu Yang, Steven Y Qian
ZUSAMMENFASSUNG
We recently reported that siRNA-knockdown of delta-5-desaturase (D5D), the rate-limiting enzyme converting upstream ω - 6 dihomo-γ-linolenic acid (DGLA) to arachidonic acid, promoted formation of the anti-cancer byproduct 8-hydroxyoctanoic acid (8-HOA) from COX-2-catalyzed DGLA peroxidation, consequently suppressing pancreatic cancer cell growth, migration and invasion. In this study, we have further investigated the anti-tumor effects of D5D-knockdown and the resulting intensified COX-2-catalyzed DGLA peroxidation in subcutaneous xenograft tumors. Four-week old female nude mice (Jackson Laboratory, J:Nu-007850) were injected with human pancreatic cancer cell line BxPC-3 or its D5D knockdown counterpart (via shRNA), followed by 4-week treatments of: vehicle control, DGLA supplementation (8 mg/mouse, twice a week), gemcitabine (30 mg/kg, twice a week), and a combination of DGLA and gemcitabine. In D5D-knockdown tumors, DGLA supplementation promoted 8-HOA formation to a threshold level (> 0.3 µg/g) and resulted in significant tumor reduction (30% vs. control). The promoted 8-HOA not only induced apoptosis associated with altered expression of Bcl-2, cleaved PARP, procaspase 3 and procaspase 9, but also suppressed the tumor metastatic potential via altering MMP-2 and E-cadherin expression. DGLA supplementation resulted in similar anti-tumor effects to those of gemcitabine in our experiments, while the combined treatment led to most significant inhibitory effect on D5D-knockdown tumor growth (70% reduction vs. control). Compared to conventional COX-2 inhibition in cancer treatment, our new strategy that takes advantage of overexpressed COX-2 in cancer cells and tumors, and of abundant ω - 6 fatty acids in the daily diet, should lead us to develop a better and safer anti-pancreatic cancer therapy for patients.