Merck
  • Metformin alleviates human cellular aging by upregulating the endoplasmic reticulum glutathione peroxidase 7.
Metformin alleviates human cellular aging by upregulating the endoplasmic reticulum glutathione peroxidase 7.
Aging cell (2018-04-17)
Jingqi Fang, Jiping Yang, Xun Wu, Gangming Zhang, Tao Li, Xi'e Wang, Hong Zhang, Chih-Chen Wang, Guang-Hui Liu, Lei Wang
ZUSAMMENFASSUNG
Metformin, an FDA-approved antidiabetic drug, has been shown to elongate lifespan in animal models. Nevertheless, the effects of metformin on human cells remain unclear. Here, we show that low-dose metformin treatment extends the lifespan of human diploid fibroblasts and mesenchymal stem cells. We report that a low dose of metformin upregulates the endoplasmic reticulum-localized glutathione peroxidase 7 (GPx7). GP×7 expression levels are decreased in senescent human cells, and GPx7 depletion results in premature cellular senescence. We also indicate that metformin increases the nuclear accumulation of nuclear factor erythroid 2-related factor 2 (Nrf2), which binds to the antioxidant response elements in the GPX7 gene promoter to induce its expression. Moreover, the metformin-Nrf2-GPx7 pathway delays aging in worms. Our study provides mechanistic insights into the beneficial effects of metformin on human cellular aging and highlights the importance of the Nrf2-GPx7 pathway in pro-longevity signaling.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Monoklonaler ANTI-FLAG® M2-Antikörper in Maus hergestellte Antikörper, 1 mg/mL, clone M2, affinity isolated antibody, buffered aqueous solution (50% glycerol, 10 mM sodium phosphate, and 150 mM NaCl, pH 7.4)
Sigma-Aldrich
Anti-Kaninchen-IgG-(Gesamtmolekül)-Peroxidase in Ziege hergestellte Antikörper, affinity isolated antibody
Sigma-Aldrich
Anti-Maus-IgG (Gesamtmolekül)-Peroxidase in Ziege hergestellte Antikörper, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Anti-α-Tubulin-Antikörper, monoklonaler Antikörper der Maus, clone B-5-1-2, purified from hybridoma cell culture
Sigma-Aldrich
Monoklonale Anti-GAPDH−-Peroxidase in Maus hergestellte Antikörper, clone GAPDH-71.1, purified immunoglobulin, buffered aqueous solution
Sigma-Aldrich
Anti-Ero1 Antibody, clone 2G4/12, clone 2G4/12, from mouse