Merck
  • Dibenzoheptazethrene isomers with different biradical characters: an exercise of Clar?s aromatic sextet rule in singlet biradicaloids.
Dibenzoheptazethrene isomers with different biradical characters: an exercise of Clar?s aromatic sextet rule in singlet biradicaloids.
Journal of the American Chemical Society (2013)
Sun, Zhe et al.