Merck
  • Atypical serum transferrin isoform distribution in liver cirrhosis studied by HPLC, capillary electrophoresis and transferrin genotyping.
Atypical serum transferrin isoform distribution in liver cirrhosis studied by HPLC, capillary electrophoresis and transferrin genotyping.
Clinica Chimica Acta; International Journal of Clinical Chemistry (2008)
Arndt T, et al.