Merck
  • AMPK-dependent phosphorylation of cingulin reversibly regulates its binding to actin filaments and microtubules.
AMPK-dependent phosphorylation of cingulin reversibly regulates its binding to actin filaments and microtubules.
Scientific reports (2018-10-21)
Tomoki Yano, Takayuki Torisawa, Kazuhiro Oiwa, Sachiko Tsukita
ZUSAMMENFASSUNG
Cytoskeletal organization is essential for the precise morphogenesis of cells, tissues, and organs. Cytoskeletons, bound to scaffolding proteins, regulate the apical junction complex (AJC), which is composed of tight and adherens junctions, and located at the apical side of epithelial cell sheets. Cingulin is a tight junction-associated protein that binds to both actin filaments and microtubules. However, how cingulin binds to microtubules and whether cingulin can bind to actin and microtubules simultaneously are unclear. Here we examined the mechanisms behind cingulin's cytoskeleton-binding properties. First, using total internal reflection fluorescence microscopy, we detected cingulin at microtubule cross points. We then found the interdomain interactions in cingulin molecules. Notably, we found that this interaction was regulated by AMPK-dependent phosphorylation and changed cingulin's conformation and binding properties to actin filaments and microtubules. Finally, we found that the AMPK-regulated cingulin properties regulated the barrier functions of epithelial cell sheets. We propose that the cellular metabolic state, which involves AMPK, can contribute to the organization and maintenance of epithelial tissues through cingulin's tight junction/cytoskeleton regulation.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
AICAR, ≥98% (HPLC), powder
Sigma-Aldrich
Indole-3-butyric acid potassium salt, suitable for plant cell culture, BioReagent