Merck
Mechanosensing by β1 integrin induces angiocrine signals for liver growth and survival.
Nature (2018-09-28)
Linda Lorenz, Jennifer Axnick, Tobias Buschmann, Carina Henning, Sofia Urner, Shentong Fang, Harri Nurmi, Nicole Eichhorst, Richard Holtmeier, Kálmán Bódis, Jong-Hee Hwang, Karsten Müssig, Daniel Eberhard, Jörg Stypmann, Oliver Kuss, Michael Roden, Kari Alitalo, Dieter Häussinger, Eckhard Lammert
ZUSAMMENFASSUNG
Angiocrine signals derived from endothelial cells are an important component of intercellular communication and have a key role in organ growth, regeneration and disease1-4. These signals have been identified and studied in multiple organs, including the liver, pancreas, lung, heart, bone, bone marrow, central nervous system, retina and some cancers1-4. Here we use the developing liver as a model organ to study angiocrine signals5,6, and show that the growth rate of the liver correlates both spatially and temporally with blood perfusion to this organ. By manipulating blood flow through the liver vasculature, we demonstrate that vessel perfusion activates β1 integrin and vascular endothelial growth factor receptor 3 (VEGFR3). Notably, both β1 integrin and VEGFR3 are strictly required for normal production of hepatocyte growth factor, survival of hepatocytes and liver growth. Ex vivo perfusion of adult mouse liver and in vitro mechanical stretching of human hepatic endothelial cells illustrate that mechanotransduction alone is sufficient to turn on angiocrine signals. When the endothelial cells are mechanically stretched, angiocrine signals trigger in vitro proliferation and survival of primary human hepatocytes. Our findings uncover a signalling pathway in vascular endothelial cells that translates blood perfusion and mechanotransduction into organ growth and maintenance.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Triton X-100, laboratory grade
Sigma-Aldrich
Duolink® In Situ PLA® Sonde Anti-Maus MINUS, Affinity purified Donkey anti-Mouse IgG (H+L)
Sigma-Aldrich
Duolink® In-situ-Detektionsreagenzien Orange
Sigma-Aldrich
Duolink® In Situ PLA® Sonde Anti-Ziege PLUS