Merck
  • Neuritin promotes neurite and spine growth in rat cerebellar granule cells via L-type calcium channel-mediated calcium influx.
Neuritin promotes neurite and spine growth in rat cerebellar granule cells via L-type calcium channel-mediated calcium influx.
Journal of neurochemistry (2018-06-20)
Qian-Ru Zhao, Jun-Mei Lu, Zhao-Yang Li, Yan-Ai Mei
ZUSAMMENFASSUNG
Neuritin is a neurotrophic factor that is activated by neural activity and neurotrophins. Its major function is to promote neurite growth and branching; however, the underlying mechanisms are not fully understood. To address this issue, this study investigated the effects of neuritin on neurite and spine growth and intracellular Ca2+ concentration in rat cerebellar granule neurons (CGNs). Incubation of CGNs for 24 h with neuritin increased neurite length and spine density; this effect was mimicked by insulin and abolished by inhibiting insulin receptor (IR) or mitogen-activated protein kinase kinase/extracellular signal-regulated kinase (ERK) activity. Calcium imaging and western blot analysis revealed that neuritin enhanced the increase in intracellular Ca2+ level induced by high K+ , and stimulated the cell surface expression of CaV 1.2 and CaV 1.3 α subunits of the L-type calcium channel, which was suppressed by inhibition of IR or mitogen-activated protein kinase kinase/ERK. Treatment with inhibitors of L-type calcium channels, calmodulin, and calcineurin (CaN) abrogated the effects of neuritin on neurite length and spine density. A similar result was obtained by silencing nuclear factor of activated T cells c4, which is known to be activated by neuritin in CGNs. These results indicate that IR and ERK signaling as well as the Ca2+ /CaN/nuclear factor of activated T cells c4 axis mediate the effects of neuritin on neurite and spine growth in CGNs. Open Science: This manuscript was awarded with the Open Materials Badge. For more information see: https://cos.io/our-services/open-science-badges/ Cover Image for this issue: doi: 10.1111/jnc.14195.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Proteasehemmer-Cocktail, for use with mammalian cell and tissue extracts, DMSO solution
Sigma-Aldrich
Poly-L-Lysin -hydrobromid, mol wt 30,000-70,000
Sigma-Aldrich
Trypsin-Inhibitor aus Glycine max (soybean), lyophilized powder
Sigma-Aldrich
Tetraethylammoniumchlorid, ≥98% (titration)
Sigma-Aldrich
4-Aminopyridin, 98%
Sigma-Aldrich
Trypsin aus Rinderpankreas, Type I, ~10,000 BAEE units/mg protein
Sigma-Aldrich
Insulin aus Rinderpankreas, Hybri-Max, powder, suitable for hybridoma
Sigma-Aldrich
U0126 ethanolate, ≥98% (HPLC), powder
Sigma-Aldrich
Calmidazoliumchlorid, solid