- TRPM7 and MagT1 in the osteogenic differentiation of human mesenchymal stem cells in vitro.
TRPM7 and MagT1 in the osteogenic differentiation of human mesenchymal stem cells in vitro.
Scientific reports (2018-11-06)
Sara Castiglioni, Valentina Romeo, Laura Locatelli, Alessandra Cazzaniga, Jeanette A M Maier
PMID30385806
ZUSAMMENFASSUNG
Mesenchymal stem cells are fundamental for bone formation and repair since they respond to microenvironmental stimuli by undergoing osteogenic differentiation. We show that the kinase and cation channel TRPM7 and the magnesium transporter MagT1 have a role in harmonizing the osteogenic differentiation of human mesenchymal stem cells. TRPM7 and MagT1 are upregulated in osteogenic differentiation and silencing either one accelerates osteogenic differentiation, partly through the activation of autophagy. Intriguingly, similar results were obtained when the cells were cultured under magnesium deficient conditions. These results underpin the contribution of magnesium, TRPM7 and MagT1 to autophagy and osteoblastogenesis.