Merck
A novel capacitive sensor based on molecularly imprinted nanoparticles as recognition elements.
Biosensors & bioelectronics (2018-09-03)
Francesco Canfarotta, Joanna Czulak, Antonio Guerreiro, Alvaro Garcia Cruz, Sergey Piletsky, Gizem Ertürk Bergdahl, Martin Hedström, Bo Mattiasson
ZUSAMMENFASSUNG
Molecularly Imprinted Polymers (MIPs) are synthetic receptors capable of selective binding to their target (template) molecules and, hence, are used as recognition elements in assays and sensors as a replacement for relatively unstable enzymes and antibodies. Herein, we describe a manufacturing-friendly protocol for integration of MIP nanoparticles (nanoMIPs) with a (label-free) capacitive sensor. The nanoMIPs were produced by solid-phase synthesis for two templates with different sizes and properties, including a small molecule tetrahydrocannabinol (THC) and a protein (trypsin). NanoMIPs were deposited on the surface of the sensor and the change in capacitance (ΔC) upon binding of the target was measured. The significant improvement in the selectivity and limit of detection (one order of magnitude compared to previously used MIP microparticles) can be attributed to their increased surface-to-volume ratio and higher specificity of the nanoMIPs produced by the solid-phase method. The methodology described is also compatible with common sensor fabrication approaches, as opposed to methods involving in situ MIP polymerisation. The proposed sensor shows high selectivity, fast sensor response (45 min including injection, regeneration and re-equilibration with running buffer), and straightforward data analysis, which makes it viable for label-free monitoring in real-time. The set of targets assessed in this manuscript shows the general applicability of the biosensor platform.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Ethylenglykoldimethacrylat, 98%, contains 90-110 ppm monomethyl ether hydroquinone as inhibitor
Sigma-Aldrich
N-(3-Aminopropyl)methacrylamid -hydrochlorid, contains ≤1,000 ppm MEHQ as stabilizer, 98% (HPLC)
Sigma-Aldrich
5-(N-Ethyl-N-isopropyl)amilorid
Sigma-Aldrich
1-[2-(Trifluoromethyl)phenyl]imidazole