Merck
  • Dual bioactivity of resveratrol fatty alcohols: differentiation of neural stem cells and modulation of neuroinflammation.
Dual bioactivity of resveratrol fatty alcohols: differentiation of neural stem cells and modulation of neuroinflammation.
Bioorganic & medicinal chemistry letters (2007-06-15)
Frédérique Hauss, Jiawei Liu, Alessandro Michelucci, Djalil Coowar, Eleonora Morga, Paul Heuschling, Bang Luu
ZUSAMMENFASSUNG
The synthesis of resveratrol fatty alcohols (RFAs), a new class of small molecules presenting strong potential for the treatment of neurological diseases, is described. RFAs, hybrid compounds combining the resveratrol nucleus and omega-alkanol side chains, are able to modulate neuroinflammation and to induce differentiation of neural stem cells into mature neurons. Acting on neuroprotection and neuroregeneration, RFAs represent an innovative approach for the treatment or cure of neuropathies.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Retinsäure, ≥98% (HPLC), powder
Sigma-Aldrich
Resveratrol, ≥99% (HPLC)
Supelco
Resveratrol, analytical standard