Merck
  • Natural molecules induce and synergize to boost expression of the human antimicrobial peptide β-defensin-3.
Natural molecules induce and synergize to boost expression of the human antimicrobial peptide β-defensin-3.
Proceedings of the National Academy of Sciences of the United States of America (2018-10-03)
Emmanuel Sechet, Erica Telford, Clément Bonamy, Philippe J Sansonetti, Brice Sperandio
ZUSAMMENFASSUNG
Antimicrobial peptides (AMPs) are mucosal defense effectors of the human innate immune response. In the intestine, AMPs are produced and secreted by epithelial cells to protect the host against pathogens and to support homeostasis with commensals. The inducible nature of AMPs suggests that potent inducers could be used to increase their endogenous expression for the prevention or treatment of diseases. Here we aimed at identifying molecules from the natural pharmacopoeia that induce expression of human β-defensin-3 (HBD3), one of the most efficient AMPs, without modifying the production of proinflammatory cytokines. By screening, we identified three molecules isolated from medicinal plants, andrographolide, oridonin, and isoliquiritigenin, which induced HBD3 production in human colonic epithelial cells. This effect was observed without activation of the NF-κB pathway or the expression of associated proinflammatory cytokines. We identified the EGF receptor as the target of these compounds and characterized the downstream-activated MAPK pathways. At the chromatin level, molecules increased phosphorylation of histone H3 on serine S10 and recruitment of the c-Fos, c-Jun, and Elk1 or c-Myc transcription factors at the HBD3 promoter. Interestingly, stimulating cells with a combination of andrographolide and isoliquiritigenin synergistically enhanced HBD3 induction 10-fold more than observed with each molecule alone. Finally, we investigated the molecular basis governing the synergistic effect, confirmed our findings in human colonic primary cells, and demonstrated that synergism increased cellular antimicrobial activity. This work shows the capability of small molecules to achieve induction of epithelial antimicrobial defenses while simultaneously avoiding the deleterious risks of an inflammatory response.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Anti-Aktin, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Anti-Acetyl-Histon-H4-Antikörper, serum, Upstate®
Sigma-Aldrich
Anti-Acetyl-Histon-H3(Lys 4)-Antikörper, Upstate®, from rabbit