Merck
Effects of allantoin and dimethyl sulfoxide on the thermal aggregation of lysozyme.
International journal of biological macromolecules (2018-07-17)
Suguru Nishinami, Atsushi Hirano, Tsutomu Arakawa, Kentaro Shiraki
ZUSAMMENFASSUNG
Allantoin is used to suppress protein aggregation without decreasing the melting temperature. However, the solubility of allantoin in water or buffer solutions is too low (approximately 30 mM at ambient temperature) to be used as a protein aggregation suppressor in various situations. Here we show that a high-concentration solution of allantoin in neat dimethyl sulfoxide (DMSO) is useful as a stock solution for the additive that controls protein aggregation. The allantoin concentration from 10 to 100 mM in 10% (v/v) DMSO significantly suppressed the thermal aggregation of hen egg white lysozyme as a model protein, with increasing allantoin concentration. The residual activity of lysozyme in 10% DMSO and 100 mM allantoin after heating at 90 °C for 10 min was increased >3-fold compared to that without allantoin. Thus, it was concluded that allantoin in DMSO is an effective stock solution for practical application in enhancing the recovery of enzymatic activity and suppressing the formation of small aggregate of protein.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Thioflavin T, used as stain for amyloid
Sigma-Aldrich
Hydantoin, 98%