Merck
  • Nicotinamide nucleotide transhydrogenase-mediated redox homeostasis promotes tumor growth and metastasis in gastric cancer.
Nicotinamide nucleotide transhydrogenase-mediated redox homeostasis promotes tumor growth and metastasis in gastric cancer.
Redox biology (2018-07-31)
Shuai Li, Zhuonan Zhuang, Teng Wu, Jie-Chun Lin, Ze-Xian Liu, Li-Fen Zhou, Ting Dai, Lei Lu, Huai-Qiang Ju
ZUSAMMENFASSUNG
Overcoming oxidative stress is a critical step for tumor growth and metastasis, however the underlying mechanisms in gastric cancer remain unclear. In this study, we found that overexpression of nicotinamide nucleotide transhydrogenase (NNT) was associated with shorter overall and disease free survival in gastric cancer. The NNT is considered a key antioxidative enzyme based on its ability to regenerate NADPH from NADH. Knockdown of NNT caused significantly NADPH reduction, induced high levels of ROS and significant cell apoptosis under oxidative stress conditions such as glucose deprival and anoikis. In vivo experiments showed that NNT promoted tumor growth, lung metastasis and peritoneal dissemination of gastric cancer. Moreover, intratumoral injection of NNT siRNA significantly suppressed gastric tumor growth in patient-derived xenograft (PDX) models. Overall, our study highlights the crucial functional roles of NNT in redox regulation and tumor progression and thus raises an important therapeutic hypothesis in gastric cancer.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Poly(2-hydroxyethylmethacrylat), BioReagent, powder, suitable for cell culture
Sigma-Aldrich
Amyloid Protein Non-Aβ Component, ≥80% (HPLC)