Merck
  • Comparison of Levosimendan and Milrinone for ECLS Weaning in Patients After Cardiac Surgery-A Retrospective Before-and-After Study.
Comparison of Levosimendan and Milrinone for ECLS Weaning in Patients After Cardiac Surgery-A Retrospective Before-and-After Study.
Journal of cardiothoracic and vascular anesthesia (2018-05-12)
Annina Jacky, Alain Rudiger, Bernard Krüger, Markus J Wilhelm, Sebastian Paal, Burkhardt Seifert, Donat R Spahn, Dominique Bettex
ZUSAMMENFASSUNG
Pharmacodynamics suggests that levosimendan might be a valuable inotrope for weaning from extracorporeal life support (ECLS). As there is a paucity of evidence regarding the effectiveness and safety of such an approach, the aim was to report the authors' experiences in ECLS weaning before and after the implementation of levosimendan in clinical practice. Retrospective before-and-after study. Cardiac intensive care unit of a university hospital. A total of 64 patients under ECLS for postcardiotomy cardiac failure, who underwent an ECLS weaning trial. Group comparisons between patients treated with levosimendan and patients treated with milrinone were made with the Mann-Whitney U test or the Pearson chi-squared test. Results are given as median (interquartile range) or numbers (percentages). Of 64 patients, 26 (41%) received levosimendan. Successful ECLS weaning was achieved in 24 (92%) and 30 patients (79%) in the levosimendan and milrinone group, respectively (p = 0.18). In the levosimendan group, fewer patients had an intra-aortic balloon pump for weaning (2 [7.7%] v 15 [40%], p = 0.008). The support with norepinephrine was similar in the levosimendan and milrinone groups at the time of ECLS removal (0.06 [0.01-0.11] v 0.07 [0.01-0.16] µg/kg/min, p = 0.64) and 24 hours later (0.06 [0.04-0.09] v 0.04 [0.00-0.09] µg/kg/min, p = 0.15). Twenty-eight days (9/26 (35%) v 14/35 (40%), p = 0.28) and 180 days (13/26 [50%] v 15/34 [44%], p = 0.80) mortalities after ECLS removal were similar in the levosimendan and the milrinone groups. Levosimendan enabled ECLS weaning without increasing norepinephrine requirements when compared to a control group receiving milrinone.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Levosimendan, ≥98% (HPLC)