Merck
  • Multipotency and cardiomyogenic potential of human adipose-derived stem cells from epicardium, pericardium, and omentum
Multipotency and cardiomyogenic potential of human adipose-derived stem cells from epicardium, pericardium, and omentum
Stem Cell Research & Therapy (2016)
Wystrychowski W, et al.