Merck
  • Mitochondrial dysfunction of in vitro grown rabbit oocytes results in preimplantation embryo arrest after activation
Mitochondrial dysfunction of in vitro grown rabbit oocytes results in preimplantation embryo arrest after activation
Journal of Reproduction and Development (2007)
Kanaya H, et al.