Merck
  • Thrombin modifies growth, proliferation and apoptosis of human colon organoids: a protease-activated receptor 1-and protease-activated receptor 4-dependent mechanism
Thrombin modifies growth, proliferation and apoptosis of human colon organoids: a protease-activated receptor 1-and protease-activated receptor 4-dependent mechanism
British Journal of Pharmacology (2018)
Sebert M, et al.