Merck
  • Structures and interactions among lysozyme proteins below the isoelectric point in presence of divalent ions
Structures and interactions among lysozyme proteins below the isoelectric point in presence of divalent ions
Chemical Physics Letters (2018)
Pandit S, et al.