Merck
  • POTRA Domains, Extracellular Lid, and Membrane Composition Modulate the Conformational Stability of the beta Barrel Assembly Factor BamA
POTRA Domains, Extracellular Lid, and Membrane Composition Modulate the Conformational Stability of the beta Barrel Assembly Factor BamA
Structure (2018)
Thoma J, et al.