Merck
  • Adipocyte model of Mycobacterium tuberculosis infection reveals differential iron availability to bacilli in the lipid rich caseous environment
Adipocyte model of Mycobacterium tuberculosis infection reveals differential iron availability to bacilli in the lipid rich caseous environment
Infection and Immunity (2018)
Nandy A, et al.