Merck
  • Obtaining information about protein secondary structures in aqueous solution using Fourier transform IR spectroscopy
Obtaining information about protein secondary structures in aqueous solution using Fourier transform IR spectroscopy
Nature Protocols (2015)
Yang H, et al.