Merck
  • Gender-dependent effect of Gpbar1 genetic deletion on the metabolic profiles of diet-induced obese mice
Gender-dependent effect of Gpbar1 genetic deletion on the metabolic profiles of diet-induced obese mice
The Journal of Endocrinology (2010)
Vassileva G, et al.