Merck
  • Quercetin protects RAW264. 7 macrophages from glucosamine-induced apoptosis and lipid accumulation via the endoplasmic reticulum stress pathway
Quercetin protects RAW264. 7 macrophages from glucosamine-induced apoptosis and lipid accumulation via the endoplasmic reticulum stress pathway
Molecular Medicine Reports (2015)
Cai X, et al.