Merck
  • Metformin decreases glucose oxidation and increases the dependency of prostate cancer cells on reductive glutamine metabolism
Metformin decreases glucose oxidation and increases the dependency of prostate cancer cells on reductive glutamine metabolism
Cancer Research (2013)
Fendt SM, et al.