Merck
  • Ca2+-induced oxidative stress in brain mitochondria treated with the respiratory chain inhibitor rotenone
Ca2+-induced oxidative stress in brain mitochondria treated with the respiratory chain inhibitor rotenone
Febs Letters (2003)
Sousa SC, et al.