Merck
  • Concanavalin A induces autophagy in hepatoma cells and has a therapeutic effect in a murine in situ hepatoma model
Concanavalin A induces autophagy in hepatoma cells and has a therapeutic effect in a murine in situ hepatoma model
Hepatology (2007)
Chang CP, et al.